This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01423568_g1
See other UBE2C GE Assays ›
Gene Symbol:
Ube2c
Gene Name:
ubiquitin-conjugating enzyme E2C
Gene Aliases:
-
Chromosome Location:
Chr.3: 161236971 - 161239371 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01423568_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106542.1 NP_001100012.1 4 - 5 532 77
GenBank mRNA FQ222472.1 - 4 - 5 523 77
FQ234128.1 - 4 - 5 526 77

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
ubiquitin-dependent protein catabolic process
negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
exit from mitosis
free ubiquitin chain polymerization
protein ubiquitination
regulation of mitotic metaphase/anaphase transition
anaphase-promoting complex-dependent catabolic process
positive regulation of exit from mitosis
protein K48-linked ubiquitination
protein K11-linked ubiquitination
ubiquitin-protein transferase activity
ligase activity
ubiquitin-like protein transferase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1019604 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 540331 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1019604 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1019604 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1019604 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1019604 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10842188 - -
Affymetrix Rat Gene 1.0 ST Array 10739356 - -
Affymetrix Rat Gene 1.0 ST Array 10842189 - -
Affymetrix Rat Gene 1.0 ST Array 10842190 - -
Affymetrix Rat Gene 1.1 ST Array 10842187 - -
Affymetrix Rat Gene 1.1 ST Array 10739355 - -
Rat Genome 230 2.0 Array 1388484_at - -
Rat Genome 230 2.0 Array 1388484_PM_at - -
Affymetrix HT Rat Focus Array 1388484_at - -
Affymetrix HT Rat Focus Array 1388484_PM_at - -
Affymetrix Rat Clariom S Assay PSR0300017187.rn.1 TC0300002296.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300017188.rn.1 TC0300002296.rn.1 RTA TC
Rat Expression Set 230 1388484_PM_at - -
Rat Expression Set 230 1388484_at - -
Affymetrix Rat Gene 1.0 ST Array 10842187 - -
Rat Genome U34 Set rc_AI103150_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21940180 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1366278 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0