This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01425804_m1
See other ARHGAP25 GE Assays ›
Gene Symbol:
Arhgap25
Gene Name:
Rho GTPase activating protein 25
Gene Aliases:
RGD1562105
Chromosome Location:
Chr.4: 119249682 - 119327822 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01425804_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109247.1 NP_001102717.1 3 - 4 720 70
XM_006236838.2 XP_006236900.1 3 - 4 2879 70
XM_006236839.2 XP_006236901.1 2 - 3 575 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0400003266.rn.1

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
positive regulation of GTPase activity
GTPase activator activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P590876 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P668582 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 355685 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 691055 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 455799 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 959868 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 697467 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 377863 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 314952 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 63107 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 411127 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 947099 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 438564 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 32567 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 657454 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 311306 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 768733 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 299368 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 20140 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 287446 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 761639 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 444015 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 917688 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 490495 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 560011 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 146696 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 67958 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 723542 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 319195 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 239945 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 913956 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 560727 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 766899 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 431032 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 923328 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 580281 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 267475 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 932299 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 156248 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 655166 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 287841 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 818479 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 432540 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 348822 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 341582 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 965249 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 456566 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 546305 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 392750 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 416860 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 901483 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 149692 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 322978 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 941297 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 202302 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 696143 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 354031 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 914882 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 18577 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 813591 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 55571 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 830725 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 157526 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 127405 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 140425 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 61904 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 149184 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 110340 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 84721 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 100914 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 23778 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 49618 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 15508 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P590876 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P668582 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P668582 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P590876 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P668582 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P590876 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P668582 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P590876 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 82706 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 47756 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 51783 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10863812 - -
Affymetrix Rat Gene 1.0 ST Array 10863810 - -
Affymetrix Rat Gene 1.0 ST Array 10863811 - -
Affymetrix Rat Gene 1.0 ST Array 10863817 - -
Affymetrix Rat Gene 1.0 ST Array 10863815 - -
Affymetrix Rat Gene 1.0 ST Array 10863814 - -
Affymetrix Rat Gene 1.0 ST Array 10863816 - -
Affymetrix Rat Gene 1.0 ST Array 10863818 - -
Affymetrix Rat Gene 1.0 ST Array 10863813 - -
Affymetrix Rat Gene 1.0 ST Array 10863809 - -
Affymetrix Rat Gene 1.0 ST Array 10863819 - -
Affymetrix Rat Gene 1.1 ST Array 10863808 - -
Rat Genome 230 2.0 Array 1388940_PM_at - -
Rat Genome 230 2.0 Array 1388940_at - -
Affymetrix HT Rat Focus Array 1388940_PM_at - -
Affymetrix HT Rat Focus Array 1388940_at - -
Affymetrix Rat Clariom S Assay PSR0400023244.rn.1 TC0400003266.rn.1 RTA TC
Rat Expression Set 230 1388940_at - -
Rat Expression Set 230 1388940_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10863808 - -
Rat Genome U34 Set rc_AA799469_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21247776 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1368068 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0