This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01426103_m1
See other NFU1 GE Assays ›
Gene Symbol:
Nfu1
Gene Name:
NFU1 iron-sulfur cluster scaffold
Gene Aliases:
Hirip5
Chromosome Location:
Chr.4: 118814284 - 118834955 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01426103_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nfu1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106606.2 NP_001100076.2 3 - 4 302 82
XM_008763063.1 XP_008761285.1 3 - 4 535 82
GenBank mRNA FM058785.1 - 3 - 4 302 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0400001328.rn.1

Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

iron-sulfur cluster assembly
iron ion binding
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0400009490.rn.1 TC0400001328.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0