This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01427911_m1
See other TWSG1 GE Assays ›
Gene Symbol:
Twsg1
Gene Name:
twisted gastrulation BMP signaling modulator 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 113699152 - 113732601 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01427911_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Twsg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108811.2 NP_001102281.1 3 - 4 483 67
XM_008767381.1 XP_008765603.1 4 - 5 614 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0900001277.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s)

ossification
mesoderm formation
salivary gland morphogenesis
hemopoiesis
cell differentiation
BMP signaling pathway
positive regulation of BMP signaling pathway
negative regulation of BMP signaling pathway
forebrain development
camera-type eye development
negative regulation of osteoblast differentiation

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P335755 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 747081 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 649496 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 170697 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 282666 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 793776 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 905649 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 775529 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 712221 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 826328 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 201671 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 45461 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 153836 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 426261 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 641690 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 590072 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 301631 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 746040 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 463667 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 211557 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 554067 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 769495 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 201236 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 162548 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 122280 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 763438 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 159046 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 152010 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 78748 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 152197 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P335755 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P335755 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P335755 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P335755 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 24036 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10930303 - -
Affymetrix Rat Gene 1.0 ST Array 10930305 - -
Affymetrix Rat Gene 1.0 ST Array 10930304 - -
Affymetrix Rat Gene 1.0 ST Array 10930302 - -
Affymetrix Rat Gene 1.0 ST Array 10930306 - -
Affymetrix Rat Gene 1.1 ST Array 10930301 - -
Rat Genome 230 2.0 Array 1397541_at - -
Rat Genome 230 2.0 Array 1376575_at - -
Rat Genome 230 2.0 Array 1397541_PM_at - -
Rat Genome 230 2.0 Array 1376575_PM_at - -
Affymetrix Rat Clariom S Assay PSR0900010860.rn.1 TC0900001277.rn.1 RTA TC
Rat Expression Set 230 1376575_at - -
Rat Expression Set 230 1376575_PM_at - -
Rat Expression Set 230 1397541_PM_at - -
Rat Expression Set 230 1397541_at - -
Affymetrix Rat Gene 1.0 ST Array 10930301 - -
Rat Genome U34 Set rc_AA926331_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21778728 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1366366 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0