This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01428854_m1
See other GSTT1 GE Assays ›
Gene Symbol:
Gstt1
Gene Name:
glutathione S-transferase theta 1
Gene Aliases:
GSTYRS
Chromosome Location:
Chr.20: 13799102 - 13816527 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01428854_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gstt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053293.2 NP_445745.1 4 - 5 554 112
XM_008772859.1 XP_008771081.1 3 - 4 750 112
XM_008772860.1 XP_008771082.1 4 - 5 1051 112
XM_008772861.1 XP_008771083.1 2 - 3 196 112
GenBank mRNA BC086426.1 - 4 - 5 554 7301492 112
FJ179408.1 - 4 - 5 524 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC2000001804.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gstt1

Gene Ontology Categories:

Function(s) Process(es)

DNA modification
glutathione metabolic process
response to salicylic acid
response to selenium ion
response to organic cyclic compound
dichloromethane metabolic process
response to vitamin E
response to drug
oxidation-reduction process
glutathione transferase activity
glutathione peroxidase activity
alkylhalidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR2000014585.rn.1 TC2000001804.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0