This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01429259_g1
See other NDUFV3 GE Assays ›
Gene Symbol:
Ndufv3
Gene Name:
NADH dehydrogenase (ubiquinone) flavoprotein 3
Gene Aliases:
Mipp65, Ndufv3l
Chromosome Location:
Chr.20: 10265826 - 10275298 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01429259_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufv3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101011.2 NP_001094481.1 1 - 2 139 89
NM_022607.2 NP_072129.2 1 - 2 139 89
GenBank mRNA AB000098.1 - 1 - 2 70 89
BC059134.1 - 95 89
CK365764.1 - 1 - 2 150 89
DV725422.1 - 1 - 2 137 89
FQ216920.1 - 1 - 2 87 89
FQ217920.1 - 1 - 2 87 89
FQ218041.1 - 1 - 2 87 89
FQ223657.1 - 1 - 2 87 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB000098.1,BC059134.1,NM_001101011.2,NM_022607.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufv3

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Process(es)

poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR2000003451.rn.1 TC2000000364.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR2000003450.rn.1 TC2000000364.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR2000003452.rn.1 TC2000000364.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-9k2pj:80/100.66.79.197:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0