Assay Details
| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012529.3 | NP_036661.3 | 274 | 80 | ||
| GenBank mRNA | BC065307.1 | - | 256 | 5598839 | 80 | |
| BC070955.1 | - | 261 | 7104205 | 80 | ||
| BC087656.1 | - | 273 | 80 | |||
| BC099814.1 | - | 258 | 7929525 | 80 | ||
| BC127477.1 | - | 264 | 8367180 | 80 | ||
| FQ213834.1 | - | 249 | 80 | |||
| M14400.1 | - | 286 | 80 | |||
| M57664.1 | - | 1 - 2 | 137 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ckb
Genomic Map