This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01429860_m1
See other PPP2R5C GE Assays ›
Gene Symbol:
Ppp2r5c
Gene Name:
protein phosphatase 2, regulatory subunit B', gamma
Gene Aliases:
-
Chromosome Location:
Chr.6: 134804134 - 134941912 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01429860_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r5c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001191112.1 NP_001178041.1 10 - 11 1230 114
XM_006240559.2 XP_006240621.1 10 - 11 1253 114
XM_008764959.1 XP_008763181.1 12 - 13 1351 114
XM_008764960.1 XP_008763182.1 12 - 13 1351 114
XM_008764961.1 XP_008763183.1 12 - 13 1351 114
XM_008764962.1 XP_008763184.1 12 - 13 1351 114
XM_008764963.1 XP_008763185.1 10 - 11 2730 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600001562.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
negative regulation of cell proliferation
protein processing
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of catalytic activity
peptidase activity
protein phosphatase type 2A regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600010742.rn.1 TC0600001562.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600010739.rn.1 TC0600001562.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600010740.rn.1 TC0600001562.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline