This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01430329_g1
See other NUDT14 GE Assays ›
Gene Symbol:
Nudt14
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 14
Gene Aliases:
-
Chromosome Location:
Chr.6: 137738155 - 137745143 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01430329_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106760.1 NP_001100230.1 3 - 4 262 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600003259.rn.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

pyrophosphatase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
UDP-sugar diphosphatase activity
metal ion binding
ADP-ribose diphosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600023019.rn.1 TC0600003259.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline