This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01431696_m1
See other COTL1 GE Assays ›
Gene Symbol:
Cotl1
Gene Name:
coactosin-like F-actin binding protein 1
Gene Aliases:
Clp
Chromosome Location:
Chr.19: 52465074 - 52499433 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01431696_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cotl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108452.1 NP_001101922.1 3 - 4 479 99
GenBank mRNA BC158746.1 - 3 - 4 458 9100871 99
EF076769.1 - 3 - 4 352 99
FQ232981.1 - 3 - 4 455 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1900001580.rn.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

defense response to fungus
actin binding
enzyme binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14228 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 439999 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 151049 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 906314 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 848888 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 701924 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 622378 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 687578 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 322502 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 396259 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 154252 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 286402 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 469687 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 667193 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 577883 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 126241 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 576209 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 507083 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 657354 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 252231 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 546182 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 273066 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 305971 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 844326 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 657207 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 57823 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 66678 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 50341 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 161396 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 96373 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 22933 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14228 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14228 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14228 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14228 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 57268 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10811454 - -
Affymetrix Rat Gene 1.0 ST Array 10811455 - -
Affymetrix Rat Gene 1.0 ST Array 10811456 - -
Affymetrix Rat Gene 1.0 ST Array 10811457 - -
Affymetrix Rat Gene 1.0 ST Array 10811453 - -
Affymetrix Rat Gene 1.1 ST Array 10811452 - -
Rat Genome 230 2.0 Array 1388596_PM_at - -
Rat Genome 230 2.0 Array 1388596_at - -
Affymetrix HT Rat Focus Array 1388596_at - -
Affymetrix HT Rat Focus Array 1388596_PM_at - -
Affymetrix Rat Clariom S Assay PSR1900012801.rn.1 TC1900001580.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900012803.rn.1 TC1900001580.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900012802.rn.1 TC1900001580.rn.1 RTA TC
Rat Expression Set 230 1388596_PM_at - -
Rat Expression Set 230 1388596_at - -
Affymetrix Rat Gene 1.0 ST Array 10811452 - -
Rat Genome U34 Set rc_AA900474_at - -
Rat Genome U34 Set rc_AI230951_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21796467 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1358946 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0