This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01431892_m1
See other WWOX GE Assays ›
Gene Symbol:
Wwox
Gene Name:
WW domain-containing oxidoreductase
Gene Aliases:
-
Chromosome Location:
Chr.19: 46761368 - 47695247 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01431892_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Wwox
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106188.1 NP_001099658.1 5 - 6 619 58
XM_006255666.2 XP_006255728.1 5 - 6 719 58
XM_006255670.2 XP_006255732.1 5 - 6 715 58
XM_006255671.2 XP_006255733.1 5 - 6 715 58
XM_006255673.2 XP_006255735.1 5 - 6 715 58
XM_008772491.1 XP_008770713.1 5 - 6 720 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1900000644.rn.1

Set Membership:

Amplicon length less than or equal to 70 Apoptosis Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
biological_process
negative regulation of Wnt signaling pathway
positive regulation of transcription from RNA polymerase II promoter
skeletal system morphogenesis
oxidation-reduction process
cellular response to transforming growth factor beta stimulus
intrinsic apoptotic signaling pathway by p53 class mediator
extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
RNA polymerase II transcription coactivator activity
molecular_function
oxidoreductase activity
enzyme binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P167804 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 652562 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 912252 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 738171 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 396343 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 434791 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 622635 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 783438 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 540679 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 434519 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 613943 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 227598 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 121861 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 498588 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 497015 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 216568 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 665753 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 164398 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 655505 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 7578 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 330889 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 195867 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 751515 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 283622 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 396787 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 950632 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 828555 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 543131 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 443445 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 194872 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 92769 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 683579 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 329468 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 761028 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 873204 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 119509 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 106899 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 776189 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 672087 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 103701 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 482785 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 195964 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 162304 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 469411 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 380470 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 437554 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 398688 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 569015 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 743890 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 708534 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 599322 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 577412 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 323508 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 4272 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 731814 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 435410 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 291649 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 113782 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 382464 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 694650 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 83810 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 772589 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 141842 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 277251 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 857784 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 416799 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 884053 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 259646 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 835496 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 135594 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 183925 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 352261 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 711855 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 85258 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 965935 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 596564 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 350487 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 385716 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 551949 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 616996 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 184319 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 146286 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 885764 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 198238 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 664224 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 443258 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 218237 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 697310 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 231950 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 627769 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 149948 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 384466 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 590943 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 109429 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 759121 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 84627 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 290441 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 686709 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 574720 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 752330 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 549785 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 581435 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 600275 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 602099 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 589395 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 209465 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 31964 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 574879 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 528993 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 242126 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 735419 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 241662 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 83094 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 23012 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 387216 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 425293 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 928757 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 59167 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 611655 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 881638 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 325023 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 427579 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 582257 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 122676 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 794008 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 99967 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 779441 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 154324 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 842042 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 152624 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 130022 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 288634 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 743807 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 459323 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 413388 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 68477 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 905697 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 533611 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 656698 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 396960 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 261521 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 327352 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 384574 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 937626 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 391708 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 639505 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 851363 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 287426 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 446131 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 580033 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 633454 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 722356 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 103321 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 826784 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 259048 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 116126 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 629079 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 408617 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 692102 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 84063 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 589053 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 412021 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 748691 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 873427 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 695660 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 849026 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 334363 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 951696 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 137729 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 141479 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 753909 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 909566 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 646453 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 520551 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 698804 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 618217 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 49080 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 612463 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 623188 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 684132 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 912723 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 249953 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 104185 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 444791 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 895160 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 887882 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 181786 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 35207 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 689894 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 532019 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 258520 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 760581 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 583107 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 368522 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 549133 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 433203 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 320405 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 302475 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 316209 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 687191 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 551406 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 672028 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 519515 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 730450 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 185097 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 875171 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 586604 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 543356 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 150328 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 765031 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 730501 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 89071 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 2100 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 159849 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 327163 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 477125 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 56272 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 795072 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 747368 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 227568 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 473161 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 7946 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 42379 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 7959 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 113782 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 144262 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 148895 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 10613 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 82815 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 107428 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 123429 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 155465 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 72963 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 112578 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 133728 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 155961 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 149054 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 87335 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 112626 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 54828 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 16880 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 68461 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 77153 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 160092 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 52139 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 129286 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 128063 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 51047 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 17038 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 35339 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 158251 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 57778 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 55529 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 68237 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 80634 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028011 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028011 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P028011 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P869415 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P513546 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 23617 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 26439 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 94760 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 92745 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 17783 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 79185 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 17238 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 89351 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 65922 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10808185 - -
Affymetrix Rat Gene 1.0 ST Array 10808186 - -
Affymetrix Rat Gene 1.0 ST Array 10808188 - -
Affymetrix Rat Gene 1.0 ST Array 10808184 - -
Affymetrix Rat Gene 1.0 ST Array 10808182 - -
Affymetrix Rat Gene 1.0 ST Array 10808183 - -
Affymetrix Rat Gene 1.0 ST Array 10808189 - -
Affymetrix Rat Gene 1.1 ST Array 10808181 - -
Rat Genome 230 2.0 Array 1396761_at - -
Rat Genome 230 2.0 Array 1391883_PM_at - -
Rat Genome 230 2.0 Array 1391883_at - -
Rat Genome 230 2.0 Array 1396761_PM_at - -
Affymetrix HT Rat Focus Array 1391883_at - -
Affymetrix HT Rat Focus Array 1391883_PM_at - -
Affymetrix Rat Clariom S Assay PSR1900005491.rn.1 TC1900000644.rn.1 RTA TC
Rat Expression Set 230 1391883_PM_at - -
Rat Expression Set 230 1396761_PM_at - -
Rat Expression Set 230 1391883_at - -
Rat Expression Set 230 1396761_at - -
Affymetrix Rat Gene 1.0 ST Array 10808181 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21129859 - -
ABI Rat Genome Survey Microarray 21861672 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1367862 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-9k2pj:80/100.66.79.197:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0