This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01431923_m1
See other ASF1B GE Assays ›
Gene Symbol:
Asf1b
Gene Name:
anti-silencing function 1B histone chaperone
Gene Aliases:
-
Chromosome Location:
Chr.19: 25077918 - 25092491 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01431923_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asf1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107160.1 NP_001100630.1 1 - 2 273 69
GenBank mRNA BC158705.1 - 1 - 2 199 9030040 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1900000310.rn.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
DNA replication-dependent nucleosome assembly
DNA replication-independent nucleosome assembly
histone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1900002344.rn.1 TC1900000310.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900002345.rn.1 TC1900000310.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900002343.rn.1 TC1900000310.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0