This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01432616_m1
See other FREM3 GE Assays ›
Gene Symbol:
Frem3
Gene Name:
FRAS1 related extracellular matrix 3
Gene Aliases:
RGD1562701
Chromosome Location:
Chr.19: 30973329 - 31031817 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01432616_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Frem3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001191699.1 NP_001178628.1 7 - 8 6147 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1900001286.rn.1

Set Membership:

Amplicon length between 86 and 100 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

Gene Ontology Categories:

Function(s)

cell communication

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P433818 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P433818 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P433818 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P433818 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P433818 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1396259_at - -
Rat Genome 230 2.0 Array 1396259_PM_at - -
Affymetrix HT Rat Focus Array 1396259_PM_at - -
Affymetrix HT Rat Focus Array 1396259_at - -
Affymetrix Rat Clariom S Assay PSR1900010338.rn.1 TC1900001286.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900010345.rn.1 TC1900001286.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900010335.rn.1 TC1900001286.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900010339.rn.1 TC1900001286.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900010340.rn.1 TC1900001286.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900010344.rn.1 TC1900001286.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900010336.rn.1 TC1900001286.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900010342.rn.1 TC1900001286.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900010343.rn.1 TC1900001286.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900010341.rn.1 TC1900001286.rn.1 RTA TC
Rat Expression Set 230 1396259_at - -
Rat Expression Set 230 1396259_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21907628 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1372981 - -
RatRef-12 Expression BeadChip ILMN_54484 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0