This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01433434_m1
See other PLA2G12B GE Assays ›
Gene Symbol:
Pla2g12b
Gene Name:
phospholipase A2, group XIIB
Gene Aliases:
-
Chromosome Location:
Chr.20: 28989440 - 29007967 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01433434_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pla2g12b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006256432.2 XP_006256494.1 2 - 3 454 85
XM_346131.7 XP_346132.5 2 - 3 453 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
cholesterol homeostasis
triglyceride homeostasis
phospholipase A2 activity
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0