Assay Details
| Gene Symbol: Swi5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246661.1 | NP_001233590.1 | 1 - 2 | 103 | 62 | |
| GenBank mRNA | BC082799.1 | - | 1 - 2 | 103 | 7304460 | 62 |
| FQ221000.1 | - | 1 - 2 | 153 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC082799.1,NM_001246661.1 transcripts.
This assay is in the 5'UTR region of the BC082799.1,NM_001246661.1 transcripts.
Set Membership: |
|

Swi5
Genomic Map