This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01435896_m1
See other GLT6D1 GE Assays ›
Gene Symbol:
Glt6d1
Gene Name:
glycosyltransferase 6 domain containing 1
Gene Aliases:
GT6m7, Gltdc1
Chromosome Location:
Chr.3: 3255645 - 3266278 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01435896_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glt6d1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106559.2 NP_001100029.2 1 - 2 85 63
XM_006233671.2 XP_006233733.1 2 - 3 205 63
XM_008761621.1 XP_008759843.1 2 - 3 104 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300002702.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
transferase activity, transferring hexosyl groups

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P445595 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 725996 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 342191 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 826302 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 316856 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 933619 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 275371 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P445595 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P445595 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P445595 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P445595 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10843602 - -
Affymetrix Rat Gene 1.0 ST Array 10843605 - -
Affymetrix Rat Gene 1.0 ST Array 10843603 - -
Affymetrix Rat Gene 1.0 ST Array 10843604 - -
Affymetrix Rat Gene 1.1 ST Array 10843600 - -
Affymetrix Rat Clariom S Assay PSR0300020261.rn.1 TC0300002702.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300020259.rn.1 TC0300002702.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300020260.rn.1 TC0300002702.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300020256.rn.1 TC0300002702.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300020257.rn.1 TC0300002702.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10843600 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21509977 - -
ABI Rat Genome Survey Microarray 21783604 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1365026 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0