This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01436121_m1
See other PPP2R4 GE Assays ›
Gene Symbol:
Ppp2r4
Gene Name:
protein phosphatase 2A activator, regulatory subunit 4
Gene Aliases:
-
Chromosome Location:
Chr.3: 8982122 - 9012585 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01436121_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108577.2 NP_001102047.1 5 - 6 641 93
GenBank mRNA BC166861.1 - 5 - 6 641 9101723 93
FQ223957.1 - 6 - 7 796 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300000212.rn.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase activator phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
metabolic process
mitotic spindle organization in nucleus
negative regulation of phosphoprotein phosphatase activity
positive regulation of phosphoprotein phosphatase activity
positive regulation of protein dephosphorylation
negative regulation of protein dephosphorylation
positive regulation of apoptotic process
regulation of phosphoprotein phosphatase activity
peptidyl-prolyl cis-trans isomerase activity
receptor binding
ATP binding
protein tyrosine phosphatase activator activity
protein phosphatase type 2A regulator activity
ATPase activity
protein homodimerization activity
protein phosphatase 2A binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0300002455.rn.1 TC0300000212.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-xvk2x:80/100.66.77.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0