This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01436124_m1
See other PPP2R4 GE Assays ›
Gene Symbol:
Ppp2r4
Gene Name:
protein phosphatase 2A activator, regulatory subunit 4
Gene Aliases:
-
Chromosome Location:
Chr.3: 8982122 - 9012585 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01436124_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108577.2 NP_001102047.1 8 - 9 972 61
GenBank mRNA BC166861.1 - 8 - 9 972 9101723 61
FQ223957.1 - 9 - 10 1127 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300000212.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase activator phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
metabolic process
mitotic spindle organization in nucleus
negative regulation of phosphoprotein phosphatase activity
positive regulation of phosphoprotein phosphatase activity
positive regulation of protein dephosphorylation
negative regulation of protein dephosphorylation
positive regulation of apoptotic process
regulation of phosphoprotein phosphatase activity
peptidyl-prolyl cis-trans isomerase activity
receptor binding
ATP binding
protein tyrosine phosphatase activator activity
protein phosphatase type 2A regulator activity
ATPase activity
protein homodimerization activity
protein phosphatase 2A binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047074 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047068 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 186424 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 2680 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 968435 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 839493 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 438053 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 594956 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 208075 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 969758 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 224681 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 667261 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 928238 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 908989 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 360274 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 797569 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 822047 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 505914 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 189872 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 26894 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 665046 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 686303 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 372635 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 804025 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 245326 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 414569 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 767577 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 89433 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 80456 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 143448 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 89626 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 166198 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 104855 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 367 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 24615 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047074 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047068 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047068 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047074 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047074 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047068 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P442161 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 101489 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 63326 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 80977 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10835247 - -
Affymetrix Rat Gene 1.0 ST Array 10835251 - -
Affymetrix Rat Gene 1.0 ST Array 10835243 - -
Affymetrix Rat Gene 1.0 ST Array 10835245 - -
Affymetrix Rat Gene 1.0 ST Array 10835246 - -
Affymetrix Rat Gene 1.0 ST Array 10835248 - -
Affymetrix Rat Gene 1.0 ST Array 10835244 - -
Affymetrix Rat Gene 1.0 ST Array 10835250 - -
Affymetrix Rat Gene 1.0 ST Array 10835249 - -
Affymetrix Rat Gene 1.1 ST Array 10835242 - -
Rat Genome 230 2.0 Array 1388338_PM_at - -
Rat Genome 230 2.0 Array 1395409_PM_at - -
Rat Genome 230 2.0 Array 1388338_at - -
Rat Genome 230 2.0 Array 1395409_at - -
Affymetrix HT Rat Focus Array 1388338_PM_at - -
Affymetrix HT Rat Focus Array 1388338_at - -
Affymetrix HT Rat Focus Array 1395409_PM_at - -
Affymetrix HT Rat Focus Array 1395409_at - -
Affymetrix Rat Clariom S Assay PSR0300002461.rn.1 TC0300000212.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300002463.rn.1 TC0300000212.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300002464.rn.1 TC0300000212.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300002465.rn.1 TC0300000212.rn.1 RTA TC
Rat Expression Set 230 1388338_at - -
Rat Expression Set 230 1388338_PM_at - -
Rat Expression Set 230 1395409_at - -
Rat Expression Set 230 1395409_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10835242 - -
Rat Genome U34 Set rc_AA849752_at - -
Rat Genome U34 Set rc_AA945759_s_at - -
Rat Genome U34 Set rc_AI012213_at - -
Rat Genome U34 Set rc_AI231775_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22317187 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1373881 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0