This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01436406_m1
See other GTF3C4 GE Assays ›
Gene Symbol:
Gtf3c4
Gene Name:
general transcription factor IIIC, polypeptide 4
Gene Aliases:
-
Chromosome Location:
Chr.3: 7404857 - 7422779 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01436406_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf3c4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109473.1 NP_001102943.1 1 - 2 458 147
XM_006233871.2 XP_006233933.1 1 - 2 445 147

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300002788.rn.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0300021224.rn.1 TC0300002788.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300021227.rn.1 TC0300002788.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300021228.rn.1 TC0300002788.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300021225.rn.1 TC0300002788.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline