This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01437280_m1
See other REV1 GE Assays ›
Gene Symbol:
Rev1
Gene Name:
REV1, polymerase (DNA directed)
Gene Aliases:
Rev1l
Chromosome Location:
Chr.9: 44714137 - 44786014 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01437280_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rev1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108213.1 NP_001101683.1 5 - 6 1221 60
XM_006244819.2 XP_006244881.1 6 - 7 1953 60
XM_006244820.2 XP_006244882.1 6 - 7 2573 60
XM_006244821.2 XP_006244883.1 5 - 6 1755 60
XM_008767030.1 XP_008765252.1 2 - 3 760 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0900001854.rn.1

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

response to UV
error-prone translesion synthesis
magnesium ion binding
damaged DNA binding
DNA-directed DNA polymerase activity
deoxycytidyl transferase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P100980 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 410337 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 819855 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 60517 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 573168 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 611510 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 904364 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 868948 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 462242 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 934762 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 400125 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 934601 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 395574 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 204755 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 413140 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 573894 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 684411 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 690112 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 397851 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 857193 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 209247 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 342548 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 743775 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 4123 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 931001 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 752490 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 687807 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 742041 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 379111 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 4429 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 440323 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 382138 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 524235 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 746103 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 115189 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 793061 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 730244 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 532239 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 115424 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 145727 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 131950 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 77433 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 143205 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P100980 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P100980 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P100980 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P100980 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 75605 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10927545 - -
Affymetrix Rat Gene 1.0 ST Array 10927556 - -
Affymetrix Rat Gene 1.0 ST Array 10927555 - -
Affymetrix Rat Gene 1.0 ST Array 10927557 - -
Affymetrix Rat Gene 1.0 ST Array 10927542 - -
Affymetrix Rat Gene 1.0 ST Array 10927558 - -
Affymetrix Rat Gene 1.0 ST Array 10927549 - -
Affymetrix Rat Gene 1.0 ST Array 10927559 - -
Affymetrix Rat Gene 1.0 ST Array 10927551 - -
Affymetrix Rat Gene 1.0 ST Array 10927552 - -
Affymetrix Rat Gene 1.0 ST Array 10927543 - -
Affymetrix Rat Gene 1.0 ST Array 10927540 - -
Affymetrix Rat Gene 1.0 ST Array 10927539 - -
Affymetrix Rat Gene 1.0 ST Array 10927546 - -
Affymetrix Rat Gene 1.0 ST Array 10927554 - -
Affymetrix Rat Gene 1.0 ST Array 10927544 - -
Affymetrix Rat Gene 1.0 ST Array 10927541 - -
Affymetrix Rat Gene 1.0 ST Array 10927548 - -
Affymetrix Rat Gene 1.0 ST Array 10927550 - -
Affymetrix Rat Gene 1.0 ST Array 10927553 - -
Affymetrix Rat Gene 1.0 ST Array 10927560 - -
Affymetrix Rat Gene 1.0 ST Array 10927547 - -
Affymetrix Rat Gene 1.1 ST Array 10927538 - -
Rat Genome 230 2.0 Array 1383347_PM_at - -
Rat Genome 230 2.0 Array 1398494_at - -
Rat Genome 230 2.0 Array 1383347_at - -
Rat Genome 230 2.0 Array 1398494_PM_at - -
Affymetrix HT Rat Focus Array 1398494_at - -
Affymetrix HT Rat Focus Array 1383347_at - -
Affymetrix HT Rat Focus Array 1398494_PM_at - -
Affymetrix HT Rat Focus Array 1383347_PM_at - -
Affymetrix Rat Clariom S Assay PSR0900015320.rn.1 TC0900001854.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900015321.rn.1 TC0900001854.rn.1 RTA TC
Rat Expression Set 230 1383347_at - -
Rat Expression Set 230 1383347_PM_at - -
Rat Expression Set 230 1398494_PM_at - -
Rat Expression Set 230 1398494_at - -
Affymetrix Rat Gene 1.0 ST Array 10927538 - -
Rat Genome U34 Set rc_AA892056_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22295907 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1350640 - -
Illumina ratRef-12 v1.0 expression beadchip ILMN_2039717 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6s775:80/100.66.79.102:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0