This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01439398_m1
See other HACD2 GE Assays ›
Gene Symbol:
Hacd2
Gene Name:
3-hydroxyacyl-CoA dehydratase 2
Gene Aliases:
Ptplb
Chromosome Location:
Chr.11: 68893684 - 68985395 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01439398_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hacd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006248438.2 XP_006248500.1 5 - 6 541 163

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Protein Phosphatase 3' Most Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

sphingolipid biosynthetic process
fatty acid elongation
very long-chain fatty acid biosynthetic process
protein tyrosine phosphatase activity
3-hydroxyacyl-CoA dehydratase activity
enzyme binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P132784 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1049289 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10754508 - -
Affymetrix Rat Gene 1.0 ST Array 10754509 - -
Affymetrix Rat Gene 1.0 ST Array 10754507 - -
Rat Genome 230 2.0 Array 1374393_at - -
Rat Genome 230 2.0 Array 1374393_PM_at - -
Affymetrix HT Rat Focus Array 1374393_PM_at - -
Affymetrix HT Rat Focus Array 1374393_at - -
Rat Expression Set 230 1374393_PM_at - -
Rat Expression Set 230 1374393_at - -
Affymetrix Rat Gene 1.0 ST Array 10754506 - -
Rat Genome U34 Set rc_AI176548_at - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1374372 - -
RatRef-12 Expression BeadChip ILMN_63938 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0