This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01439403_m1
See other HACD2 GE Assays ›
Gene Symbol:
Hacd2
Gene Name:
3-hydroxyacyl-CoA dehydratase 2
Gene Aliases:
Ptplb
Chromosome Location:
Chr.11: 68893684 - 68985395 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01439403_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hacd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006248438.2 XP_006248500.1 2 - 3 302 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_006248438.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Protein Phosphatase Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

sphingolipid biosynthetic process
fatty acid elongation
very long-chain fatty acid biosynthetic process
protein tyrosine phosphatase activity
3-hydroxyacyl-CoA dehydratase activity
enzyme binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0