This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01439427_m1
See other IGSF11 GE Assays ›
Gene Symbol:
Igsf11
Gene Name:
immunoglobulin superfamily, member 11
Gene Aliases:
-
Chromosome Location:
Chr.11: 64286691 - 64421248 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01439427_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013120.1 NP_001013138.1 6 - 7 1046 150
GenBank mRNA BC085929.1 - 6 - 7 1046 7110445 150

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100001644.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igsf11

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin

Gene Ontology Categories:

Function(s)

cell adhesion
regulation of growth

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P648707 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P218305 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 628141 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 85145 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 265120 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 446854 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 566904 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 240600 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 255038 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 638104 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 587729 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 816305 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 618444 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 172433 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 687695 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 117183 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 467814 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 889593 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 817831 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 39035 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 260704 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 435483 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 633882 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 19137 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 374560 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 514307 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 746016 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 572350 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 808151 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 138758 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 598379 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 874376 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 519668 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 111144 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 67205 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 784186 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 692023 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 238656 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 889373 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 202744 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 922154 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 447070 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 897618 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 409427 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 839485 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 937678 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 950616 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 717718 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 426851 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 465138 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 860050 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 822702 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 354126 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 418721 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 403186 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 923091 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 608326 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 857644 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 504159 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 74249 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 339963 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 722268 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 565614 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 793668 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 209344 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 202187 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 934345 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 757031 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 178752 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 713469 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 49378 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 968523 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 221993 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 671581 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 871204 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 532915 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 856200 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 573991 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 9792 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 187971 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 15143 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 740698 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 883310 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 507414 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 544519 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 587537 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 820356 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 190641 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 445074 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 895036 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 323196 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 500776 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 664057 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 204760 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 406943 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 74754 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 27468 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 66736 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 50334 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 46273 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 120712 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 102478 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 24122 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 33587 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 163114 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 37148 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 123507 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 130826 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 11719 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 129778 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 33134 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P218305 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P648707 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P218305 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P648707 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P218305 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P648707 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P218305 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 69113 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 78841 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 6944 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 68403 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 87565 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 76255 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 101027 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 29399 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 48950 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 12713 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 7540 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10754153 - -
Affymetrix Rat Gene 1.0 ST Array 10754150 - -
Affymetrix Rat Gene 1.0 ST Array 10754148 - -
Affymetrix Rat Gene 1.0 ST Array 10754147 - -
Affymetrix Rat Gene 1.0 ST Array 10754146 - -
Affymetrix Rat Gene 1.0 ST Array 10754149 - -
Affymetrix Rat Gene 1.0 ST Array 10754151 - -
Affymetrix Rat Gene 1.0 ST Array 10754152 - -
Affymetrix Rat Gene 1.1 ST Array 10754145 - -
Rat Genome 230 2.0 Array 1376733_PM_at - -
Rat Genome 230 2.0 Array 1376733_at - -
Affymetrix HT Rat Focus Array 1376733_at - -
Affymetrix HT Rat Focus Array 1376733_PM_at - -
Affymetrix Rat Clariom S Assay PSR1100009906.rn.1 TC1100001644.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100009907.rn.1 TC1100001644.rn.1 RTA TC
Rat Expression Set 230 1376733_at - -
Rat Expression Set 230 1376733_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10754145 - -
Rat Genome U34 Set rc_AI072647_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21798000 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1650776 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0