This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01439490_m1
See other MOB3B GE Assays ›
Gene Symbol:
Mob3b
Gene Name:
MOB kinase activator 3B
Gene Aliases:
Mobkl2b, RGD1560696
Chromosome Location:
Chr.5: 50437444 - 50638157 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01439490_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mob3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108970.1 NP_001102440.1 2 - 3 834 101
XM_006238001.2 XP_006238063.1 2 - 3 847 101
XM_008763630.1 XP_008761852.1 2 - 3 1525 101
XM_008763631.1 XP_008761853.1 4 - 5 1459 101
XM_008763632.1 XP_008761854.1 3 - 4 1353 101
XM_008763633.1 XP_008761855.1 2 - 3 1106 101
XM_008763634.1 XP_008761856.1 2 - 3 855 101
XM_008763635.1 XP_008761857.1 4 - 5 1878 101
XM_008763636.1 XP_008761858.1 2 - 3 1523 101
GenBank mRNA FQ220583.1 - 1 - 2 325 101
FQ222144.1 - 2 - 3 792 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500002504.rn.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

kinase activator

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0500018543.rn.1 TC0500002504.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0