This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01439508_m1
See other SMIM8 GE Assays ›
Gene Symbol:
Smim8
Gene Name:
small integral membrane protein 8
Gene Aliases:
RGD1305158
Chromosome Location:
Chr.5: 50230617 - 50239647 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01439508_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smim8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106644.1 NP_001100114.1 1 - 2 305 61

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001106644.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0500018498.rn.1 TC0500002498.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500018510.rn.1 TC0500002498.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0