This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01440023_mH
See other GABRR1 GE Assays ›
Gene Symbol:
Gabrr1
Gene Name:
gamma-aminobutyric acid (GABA) A receptor, rho 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 48345279 - 48411170 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01440023_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gabrr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017291.3 NP_058987.2 9 - 10 1260 134
XM_008763609.1 XP_008761831.1 9 - 10 1043 134
GenBank mRNA U21070.1 - 9 - 10 1217 134
X95579.1 - 9 - 10 1260 134

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500000404.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel 3' Most Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gabrr1

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
visual perception
ion transmembrane transport
chloride transmembrane transport
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
protein binding
GABA receptor activity
protein domain specific binding
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017148 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 961550 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 312835 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 63658 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 687254 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 962680 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 33121 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 425998 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 69751 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 36477 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 571544 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 439178 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 872870 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 439311 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 739196 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 469392 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 857291 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 219580 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 146163 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 839223 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 151024 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 875961 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 235462 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 84208 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 163359 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 131454 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 114367 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 26976 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017148 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017148 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017148 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11898 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 35943 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 99812 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 81211 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10868154 - -
Affymetrix Rat Gene 1.0 ST Array 10868152 - -
Affymetrix Rat Gene 1.0 ST Array 10868158 - -
Affymetrix Rat Gene 1.0 ST Array 10868156 - -
Affymetrix Rat Gene 1.0 ST Array 10868155 - -
Affymetrix Rat Gene 1.0 ST Array 10868153 - -
Affymetrix Rat Gene 1.0 ST Array 10868161 - -
Affymetrix Rat Gene 1.0 ST Array 10868160 - -
Affymetrix Rat Gene 1.0 ST Array 10868157 - -
Affymetrix Rat Gene 1.0 ST Array 10868162 - -
Affymetrix Rat Gene 1.0 ST Array 10868159 - -
Affymetrix Rat Gene 1.1 ST Array 10868151 - -
Rat Genome 230 2.0 Array 1368457_at - -
Rat Genome 230 2.0 Array 1368457_PM_at - -
Rat Genome 230 2.0 Array 1368456_at - -
Rat Genome 230 2.0 Array 1368456_PM_at - -
Affymetrix HT Rat Focus Array 1368456_PM_at - -
Affymetrix HT Rat Focus Array 1368457_at - -
Affymetrix HT Rat Focus Array 1368457_PM_at - -
Affymetrix HT Rat Focus Array 1368456_at - -
Affymetrix Rat Clariom S Assay PSR0500002517.rn.1 TC0500000404.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500002518.rn.1 TC0500000404.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500002516.rn.1 TC0500000404.rn.1 RTA TC
Rat Expression Set 230 1368456_PM_at - -
Rat Expression Set 230 1368456_at - -
Rat Expression Set 230 1368457_at - -
Rat Expression Set 230 1368457_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10868151 - -
Rat Genome U34 Set X95579_at - -
Rat Genome U34 Set X95579_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20939395 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1373838 - -
RatRef-12 Expression BeadChip ILMN_62256 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline