This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01440264_m1
See other CNTFR GE Assays ›
Gene Symbol:
Cntfr
Gene Name:
ciliary neurotrophic factor receptor
Gene Aliases:
-
Chromosome Location:
Chr.5: 58041631 - 58070125 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01440264_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cntfr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003929.1 NP_001003929.1 3 - 4 400 101
XM_006238055.2 XP_006238117.1 2 - 3 322 101
GenBank mRNA S54212.1 - 3 - 4 400 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500002579.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cntfr

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

suckling behavior
brainstem development
sex differentiation
positive regulation of cell proliferation
negative regulation of neuron apoptotic process
skeletal muscle organ development
ciliary neurotrophic factor-mediated signaling pathway
ciliary neurotrophic factor receptor activity
receptor binding
cytokine binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P103174 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103174 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103174 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P103174 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15815 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 702079664 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 18912 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 36587 - -
Agilent Rat cDNA Microarray (G4105A) 702079664 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10882375 - -
Affymetrix Rat Gene 1.0 ST Array 10882376 - -
Affymetrix Rat Gene 1.0 ST Array 10882370 - -
Affymetrix Rat Gene 1.0 ST Array 10882368 - -
Affymetrix Rat Gene 1.0 ST Array 10882371 - -
Affymetrix Rat Gene 1.0 ST Array 10882373 - -
Affymetrix Rat Gene 1.0 ST Array 10882372 - -
Affymetrix Rat Gene 1.0 ST Array 10882374 - -
Affymetrix Rat Gene 1.0 ST Array 10882369 - -
Affymetrix Rat Gene 1.1 ST Array 10882367 - -
Rat Genome 230 2.0 Array 1394008_PM_x_at - -
Rat Genome 230 2.0 Array 1392749_at - -
Rat Genome 230 2.0 Array 1394008_x_at - -
Rat Genome 230 2.0 Array 1392749_PM_at - -
Affymetrix HT Rat Focus Array 1394008_x_at - -
Affymetrix HT Rat Focus Array 1392749_at - -
Affymetrix HT Rat Focus Array 1392749_PM_at - -
Affymetrix HT Rat Focus Array 1394008_PM_x_at - -
Affymetrix Rat Clariom S Assay PSR0500019032.rn.1 TC0500002579.rn.1 RTA TC
Rat Expression Set 230 1394008_PM_x_at - -
Rat Expression Set 230 1392749_PM_at - -
Rat Expression Set 230 1394008_x_at - -
Rat Expression Set 230 1392749_at - -
Affymetrix Rat Gene 1.0 ST Array 10882367 - -
Rat Genome U34 Set S54212_at - -
Rat Genome U34 Set S54212_at - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1372239 - -
RatRef-12 Expression BeadChip ILMN_60678 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0