This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01440272_m1
See other RGD1562865 GE Assays ›
Gene Symbol:
RGD1562865
Gene Name:
similar to BTB and CNC homology 1, basic leucine zipper transcription factor 2
Gene Aliases:
Bach2
Chromosome Location:
Chr.5: 47546014 - 47807176 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01440272_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RGD1562865
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001135754.1 NP_001129226.1 5 - 6 2300 61
XM_008763617.1 XP_008761839.1 4 - 5 2393 61
XM_008763618.1 XP_008761840.1 5 - 6 2382 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500000376.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067968 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 184388 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 516901 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 97828 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 703122 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 344891 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 965842 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 214014 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 416107 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 46406 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 526427 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 642477 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 17760 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 174787 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 108511 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 597474 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 901097 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 916666 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 140488 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 264043 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 101516 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 743661 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 197654 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 110218 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 420566 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 915056 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 24472 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 391876 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 505925 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 514769 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 353901 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 605997 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 934820 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 120173 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 155180 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 747716 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 469093 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 534395 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 327484 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 663141 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 410860 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 558067 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 664058 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 202179 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 814344 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 852323 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 813810 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 773214 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 945785 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 41705 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 530511 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 743777 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 590252 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 330614 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 245004 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 822064 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 520917 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 121636 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 893527 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 424286 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 43412 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 921379 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 318315 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 684015 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 459213 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 349202 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 134989 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 379849 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 806983 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 728862 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 672980 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 731552 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 171125 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 850636 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 94994 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 528406 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 404962 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 135020 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 647336 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 888853 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 807629 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 280683 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 790879 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 371171 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 237090 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 693113 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 718496 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 387332 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 833452 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 428109 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 447036 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 493512 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 579297 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 218662 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 190750 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 214838 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 829875 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 38997 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 716605 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 269236 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 699985 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 763267 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 253380 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 866657 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 250502 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 515800 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 922979 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 822916 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 297223 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 450424 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 769120 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 720356 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 181536 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 59274 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 798538 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 380683 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 61462 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 649490 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 171309 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 712664 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 506461 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 872480 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 726921 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 155001 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 289685 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 400644 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 771664 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 568488 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 846942 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 119548 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 140067 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 227468 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 110055 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 198556 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 461319 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 290814 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 569994 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 410727 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 780275 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 611811 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 120816 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 499078 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 12716 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 225291 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 558259 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 263097 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 162084 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 130166 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 134978 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 640638 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 175655 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 911339 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 657469 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 165717 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 507491 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 185925 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 355838 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 469107 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 466766 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 168736 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 115270 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 126958 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 859415 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 298331 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 263966 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 193340 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 317414 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 357481 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 532842 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 84992 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 668547 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 488481 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 104772 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 425032 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 635092 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 615975 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 672749 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 477207 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 449135 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 754036 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 804926 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 938230 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 129401 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 949460 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 186964 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 455846 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 623455 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 351567 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 865800 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 137418 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 886841 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 446 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 161110 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 44839 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 98596 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 17838 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 163650 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 48221 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 168115 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 81253 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 3095 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 126352 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 7366 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 170602 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 36430 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 80650 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 9266 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 145833 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 171003 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 17906 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 174785 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 144097 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 170198 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 155786 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 168742 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 49541 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 157619 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 153509 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 15490 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 20542 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067968 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067968 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067968 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P410288 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10868004 - -
Affymetrix Rat Gene 1.0 ST Array 10868001 - -
Affymetrix Rat Gene 1.0 ST Array 10868002 - -
Affymetrix Rat Gene 1.0 ST Array 10868006 - -
Affymetrix Rat Gene 1.0 ST Array 10868003 - -
Affymetrix Rat Gene 1.0 ST Array 10868005 - -
Affymetrix Rat Gene 1.1 ST Array 10868000 - -
Affymetrix Rat Clariom S Assay PSR0500002270.rn.1 TC0500000376.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500002269.rn.1 TC0500000376.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10868000 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21715704 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1350062 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0