This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01440354_g1
See other NUDT2 GE Assays ›
Gene Symbol:
Nudt2
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 2
Gene Aliases:
-
Chromosome Location:
Chr.5: 57845819 - 57860658 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01440354_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207596.1 NP_997479.1 1 - 2 39 77
GenBank mRNA BC058156.1 - 1 - 2 39 6888562 77
FQ227408.1 - 1 - 2 40 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC058156.1,NM_207596.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nudt2

Panther Classification:

Molecular Function -

hydrolase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
bis(5'-nucleosyl)-tetraphosphatase (asymmetrical) activity
GTP binding
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0500002952.rn.1 TC0500000491.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500002953.rn.1 TC0500000491.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0