This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01441205_g1
See other CD99 GE Assays ›
Gene Symbol:
Cd99
Gene Name:
CD99 molecule
Gene Aliases:
-
Chromosome Location:
Chr.20: 56192426 - 56196961 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01441205_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd99
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100804.1 NP_001094274.1 8 - 9 488 81
GenBank mRNA AY262357.1 - 8 - 9 477 81
BC168196.1 - 488 7111460 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC2000001795.rn.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P153070 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153070 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153070 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P153070 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1371918_PM_at - -
Rat Genome 230 2.0 Array 1371918_at - -
Affymetrix HT Rat Focus Array 1371918_PM_at - -
Affymetrix HT Rat Focus Array 1371918_at - -
Affymetrix Rat Clariom S Assay PSR2000014307.rn.1 TC2000001795.rn.1 RTA TC
Rat Expression Set 230 1371918_at - -
Rat Expression Set 230 1371918_PM_at - -
Rat Genome U34 Set rc_AI235284_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21149466 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1530273 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline