This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01442557_m1
See other PHTF2 GE Assays ›
Gene Symbol:
Phtf2
Gene Name:
putative homeodomain transcription factor 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 10823258 - 10938273 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01442557_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phtf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106577.1 NP_001100047.1 4 - 5 326 76
XM_008762640.1 XP_008760862.1 4 - 5 281 76
XM_008762641.1 XP_008760863.1 3 - 4 1864 76
XM_008762642.1 XP_008760864.1 4 - 5 1876 76
XM_008762643.1 XP_008760865.1 3 - 4 178 76
XM_008762644.1 XP_008760866.1 4 - 5 1876 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0400000169.rn.1

Additional Information:

This assay is in the 5'UTR region of the XM_008762643.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Process(es)

DNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0400001349.rn.1 TC0400000169.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21025991 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0