Assay Details
| Gene Symbol: Pcmtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763511.1 | XP_008761733.1 | 5 - 6 | 1905 | 134 | |
| XM_008763512.1 | XP_008761734.1 | 4 - 5 | 1795 | 134 | ||
More Information
Set Membership: |
|

Pcmtd1
Genomic Map