This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01443649_m1
See other DHX15 GE Assays ›
Gene Symbol:
Dhx15 LOC100910750
Gene Name:
DEAH (Asp-Glu-Ala-His) box helicase 15
putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15-like
Gene Aliases:
-
-
Chromosome Location:
Chr.14: 61136742 - 61174880 on Build Rnor_6.0
Chromosome Location:
Chr.14: 61650943 - 61668505 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01443649_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dhx15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001191597.1 NP_001178526.1 6 - 7 1418 114
XM_006251031.2 XP_006251093.1 6 - 7 1422 114
Gene Symbol: LOC100910750
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003751372.3 XP_003751420.2 916 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1400000639.rn.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

RNA helicase

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
double-stranded RNA binding
ATP-dependent RNA helicase activity
ATP binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1400004552.rn.1 TC1400000639.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline