This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01443674_g1
See other TMED4 GE Assays ›
Gene Symbol:
Tmed4
Gene Name:
transmembrane emp24 protein transport domain containing 4
Gene Aliases:
RGD1306319
Chromosome Location:
Chr.14: 86390327 - 86395149 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01443674_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmed4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107238.1 NP_001100708.1 1 - 2 473 99
XM_006251444.2 XP_006251506.1 2 - 3 311 99
XM_006251445.2 XP_006251507.1 2 - 3 312 99
GenBank mRNA BC168726.1 - 2 - 3 283 6889842 99
BC168735.1 - 2 - 3 283 6891034 99
BC168766.1 - 2 - 3 283 6890852 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1400002191.rn.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

transport
signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
signal transducer activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1400016339.rn.1 TC1400002191.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400016337.rn.1 TC1400002191.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0