This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01443950_m1
See other NXPE3 GE Assays ›
Gene Symbol:
Nxpe3
Gene Name:
neurexophilin and PC-esterase domain family, member 3
Gene Aliases:
Fam55c
Chromosome Location:
Chr.11: 47188349 - 47238372 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01443950_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nxpe3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109435.1 NP_001102905.1 5 - 6 1611 86
XM_006248252.2 XP_006248314.1 5 - 6 1437 86
XM_006248253.2 XP_006248315.1 5 - 6 1448 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100000490.rn.1

Set Membership:

Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P885445 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P321686 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 116772 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 971948 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 152275 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 5874 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 955629 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 905088 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 344300 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 727121 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 110763 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 312793 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 677075 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 374354 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 129597 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 891386 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 435901 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 53476 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 57689 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 686402 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 895796 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 536599 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 925504 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 870697 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 631659 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 853673 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 717077 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 400817 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 395013 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 430400 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 659531 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 105426 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 193319 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 690420 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 544405 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 28687 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 88885 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 6375 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 85003 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 15155 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 166660 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P885445 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321686 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321686 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P885445 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P321686 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P885445 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P321686 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P885445 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 39600 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10750846 - -
Affymetrix Rat Gene 1.0 ST Array 10750829 - -
Affymetrix Rat Gene 1.0 ST Array 10750828 - -
Affymetrix Rat Gene 1.0 ST Array 10750832 - -
Affymetrix Rat Gene 1.0 ST Array 10750830 - -
Affymetrix Rat Gene 1.0 ST Array 10750826 - -
Affymetrix Rat Gene 1.0 ST Array 10750845 - -
Affymetrix Rat Gene 1.0 ST Array 10750840 - -
Affymetrix Rat Gene 1.0 ST Array 10750847 - -
Affymetrix Rat Gene 1.0 ST Array 10750844 - -
Affymetrix Rat Gene 1.0 ST Array 10750831 - -
Affymetrix Rat Gene 1.1 ST Array 10750825 - -
Rat Genome 230 2.0 Array 1385046_PM_at - -
Rat Genome 230 2.0 Array 1385046_at - -
Affymetrix Rat Clariom S Assay PSR1100002513.rn.1 TC1100000490.rn.1 RTA TC
Rat Expression Set 230 1385046_at - -
Rat Expression Set 230 1385046_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10750825 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21921836 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0