This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01443951_m1
See other NXPE3 GE Assays ›
Gene Symbol:
Nxpe3
Gene Name:
neurexophilin and PC-esterase domain family, member 3
Gene Aliases:
Fam55c
Chromosome Location:
Chr.11: 47188349 - 47238372 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01443951_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nxpe3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109435.1 NP_001102905.1 1 - 2 356 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001109435.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1100002506.rn.1 TC1100000490.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100002508.rn.1 TC1100000490.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100002505.rn.1 TC1100000490.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline