This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01444436_m1
See other GSTO1 GE Assays ›
Gene Symbol:
Gsto1
Gene Name:
glutathione S-transferase omega 1
Gene Aliases:
SPG-R
Chromosome Location:
Chr.1: 267607437 - 267617387 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01444436_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gsto1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007602.1 NP_001007603.1 2 - 3 224 97
GenBank mRNA AB008807.1 - 2 - 3 206 97
BC079363.1 - 2 - 3 224 7109248 97
FJ179405.1 - 2 - 3 142 97
FQ218105.1 - 2 - 3 199 97
FQ220742.1 - 2 - 3 166 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100008077.rn.1

Additional Information:

This assay is in the 5'UTR region of the AB008807.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gsto1

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
L-ascorbic acid metabolic process
L-ascorbic acid biosynthetic process
xenobiotic catabolic process
oxidation-reduction process
negative regulation of ryanodine-sensitive calcium-release channel activity
positive regulation of ryanodine-sensitive calcium-release channel activity
cellular response to arsenic-containing substance
glutathione transferase activity
oxidoreductase activity
glutathione dehydrogenase (ascorbate) activity
methylarsonate reductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100066020.rn.1 TC0100008077.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0