This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01444851_m1
See other PPIB GE Assays ›
Gene Symbol:
Ppib
Gene Name:
peptidylprolyl isomerase B
Gene Aliases:
CypB, Scylp
Chromosome Location:
Chr.8: 71719681 - 71725538 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01444851_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppib
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022536.2 NP_071981.1 1 - 2 131 180
GenBank mRNA AF071225.1 - 1 - 2 131 180
BC061971.1 - 1 - 2 160 5623342 180
FQ211358.1 - 1 - 2 135 180
FQ212031.1 - 1 - 2 160 180
FQ219792.1 - 1 - 2 169 180
FQ219926.1 - 1 - 2 181 180
FQ221020.1 - 1 - 2 184 180
FQ221248.1 - 1 - 2 179 180
FQ222696.1 - 1 - 2 293 180
FQ228087.1 - 1 - 2 174 180
FQ229174.1 - 1 - 2 181 180
FQ229214.1 - 1 - 2 175 180
FQ230004.1 - 185 180
FQ231656.1 - 1 - 2 174 180
FQ232077.1 - 1 - 2 170 180
FQ232920.1 - 1 - 2 184 180

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800000960.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppib

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
positive regulation of multicellular organism growth
positive regulation by host of viral process
positive regulation by host of viral genome replication
protein stabilization
nuclear transport
bone development
chaperone-mediated protein folding
regulation of post-translational protein modification
peptidyl-prolyl cis-trans isomerase activity
protein binding
protein complex binding
peptide binding
poly(A) RNA binding
RNA polymerase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0800006986.rn.1 TC0800000960.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800006988.rn.1 TC0800000960.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800006985.rn.1 TC0800000960.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline