This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01445039_g1
See other NUDT16 GE Assays ›
Gene Symbol:
Nudt16
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 16
Gene Aliases:
RGD1311387
Chromosome Location:
Chr.8: 113673069 - 113675124 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01445039_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127554.1 NP_001121026.1 1 - 2 141 73
GenBank mRNA BC166915.1 - 1 - 2 141 6917859 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800003294.rn.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

adenosine to inosine editing
mRNA catabolic process
proteolysis
positive regulation of cell proliferation
snoRNA catabolic process
dephosphorylation
dITP catabolic process
IDP catabolic process
chromosome organization
positive regulation of cell cycle process
XDP catabolic process
positive regulation of double-strand break repair
magnesium ion binding
mRNA binding
GTP binding
metalloexopeptidase activity
manganese ion binding
snoRNA binding
dITP diphosphatase activity
protein homodimerization activity
m7G(5')pppN diphosphatase activity
cobalt ion binding
inosine-diphosphatase activity
dIDP diphosphatase activity
nucleotide phosphatase activity, acting on free nucleotides
XTP binding
ITP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0800026290.rn.1 TC0800003294.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800026288.rn.1 TC0800003294.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline