This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01445633_m1
See other MANF GE Assays ›
Gene Symbol:
Manf
Gene Name:
mesencephalic astrocyte-derived neurotrophic factor
Gene Aliases:
Armet
Chromosome Location:
Chr.8: 115623170 - 115626408 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01445633_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Manf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108183.1 NP_001101653.1 2 - 3 272 60
XM_006243775.1 XP_006243837.1 2 - 3 267 60
GenBank mRNA BC166980.1 - 2 - 3 244 7318164 60
FQ232064.1 - 2 - 3 402 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800003331.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

vasoconstriction of artery involved in ischemic response to lowering of systemic arterial blood pressure
response to unfolded protein
biological_process
molecular_function
growth factor activity
poly(A) RNA binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P074332 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 84385 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 182050 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074332 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074332 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P074332 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P116172 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P116176 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10919999 - -
Affymetrix Rat Gene 1.0 ST Array 10919998 - -
Affymetrix Rat Gene 1.0 ST Array 10920000 - -
Affymetrix Rat Gene 1.0 ST Array 10919997 - -
Affymetrix Rat Gene 1.1 ST Array 10919996 - -
Rat Genome 230 2.0 Array 1372352_PM_at - -
Rat Genome 230 2.0 Array 1372352_at - -
Affymetrix HT Rat Focus Array 1372352_at - -
Affymetrix HT Rat Focus Array 1372352_PM_at - -
Affymetrix Rat Clariom S Assay PSR0800026849.rn.1 TC0800003331.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800026847.rn.1 TC0800003331.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800026850.rn.1 TC0800003331.rn.1 RTA TC
Rat Expression Set 230 1372352_at - -
Rat Expression Set 230 1372352_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10919996 - -
Rat Genome U34 Set rc_AI170666_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21577456 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1370585 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline