This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01446893_m1
See other UBE3D GE Assays ›
Gene Symbol:
Ube3d
Gene Name:
ubiquitin protein ligase E3D
Gene Aliases:
RGD1306773, Ube2cbp
Chromosome Location:
Chr.8: 93948689 - 94120476 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01446893_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube3d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039610.1 NP_001034699.1 1 - 2 200 84
XM_006243479.2 XP_006243541.1 1 - 2 218 84
GenBank mRNA BC101916.1 - 1 - 2 200 7935691 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800003053.rn.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0800024510.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024511.rn.1 TC0800003053.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0