This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01446894_m1
See other UBE3D GE Assays ›
Gene Symbol:
Ube3d
Gene Name:
ubiquitin protein ligase E3D
Gene Aliases:
RGD1306773, Ube2cbp
Chromosome Location:
Chr.8: 93948689 - 94120476 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01446894_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube3d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039610.1 NP_001034699.1 2 - 3 401 59
XM_006243479.2 XP_006243541.1 2 - 3 419 59
GenBank mRNA BC101916.1 - 2 - 3 401 7935691 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800003053.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
ligase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P149839 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 176054 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 939354 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 397360 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 108134 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 937010 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 342737 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 246168 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 737076 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 468948 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 434779 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 663102 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 502578 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 932174 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 868898 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 542346 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 156591 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 567242 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 750392 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 740416 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 143889 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 743681 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 773538 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 958714 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 682957 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 230980 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 783169 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 283338 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 127625 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 187548 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 391407 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 783809 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 570902 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 299561 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 656098 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 57734 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 257824 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 158534 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 800114 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 871262 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 16429 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 189292 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 465431 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 83671 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 939272 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 469087 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 638207 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 55138 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 528791 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 692743 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 959990 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 651891 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 269658 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 698498 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 784578 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 959929 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 767044 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 110855 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 277475 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 469146 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 882308 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 752265 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 404688 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 388083 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 958651 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 641447 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 625680 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 694038 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 634654 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 36242 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 189974 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 660270 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 436675 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 962159 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 966985 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 557902 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 113431 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 856062 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 920390 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 445499 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 440730 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 858988 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 598582 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 32102 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 201076 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 512778 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 249769 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 209086 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 521387 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 699028 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 913730 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 495620 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 323312 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 967992 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 627867 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 909008 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 753352 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 290007 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 798277 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 716211 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 657713 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 201061 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 2451 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 224023 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 326141 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 547002 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 104261 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 116391 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 88549 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 84175 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 41086 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 141820 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 101450 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 169580 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 129714 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 101743 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 109114 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 179088 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 164331 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 13282 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 68919 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 168452 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 50560 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 67334 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P149839 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P149839 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P149839 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19216 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 5104 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 13543 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 104086 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 57764 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 96366 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 53140 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 26367 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 2161 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 84114 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 99355 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 48772 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 84719 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10919081 - -
Affymetrix Rat Gene 1.0 ST Array 10919083 - -
Affymetrix Rat Gene 1.0 ST Array 10919088 - -
Affymetrix Rat Gene 1.0 ST Array 10919086 - -
Affymetrix Rat Gene 1.0 ST Array 10919077 - -
Affymetrix Rat Gene 1.0 ST Array 10919089 - -
Affymetrix Rat Gene 1.0 ST Array 10919085 - -
Affymetrix Rat Gene 1.0 ST Array 10919082 - -
Affymetrix Rat Gene 1.0 ST Array 10919079 - -
Affymetrix Rat Gene 1.0 ST Array 10919087 - -
Affymetrix Rat Gene 1.0 ST Array 10919078 - -
Affymetrix Rat Gene 1.0 ST Array 10919080 - -
Affymetrix Rat Gene 1.1 ST Array 10919076 - -
Rat Genome 230 2.0 Array 1392685_at - -
Rat Genome 230 2.0 Array 1392685_PM_at - -
Affymetrix HT Rat Focus Array 1392685_at - -
Affymetrix HT Rat Focus Array 1392685_PM_at - -
Affymetrix Rat Clariom S Assay PSR0800024498.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024508.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024497.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024504.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024503.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024505.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024502.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024509.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024500.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024495.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024496.rn.1 TC0800003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800024506.rn.1 TC0800003053.rn.1 RTA TC
Rat Expression Set 230 1392685_PM_at - -
Rat Expression Set 230 1392685_at - -
Affymetrix Rat Gene 1.0 ST Array 10919076 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1360263 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0