This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01448619_m1
See other PCBD2 GE Assays ›
Gene Symbol:
Pcbd2
Gene Name:
pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1) 2
Gene Aliases:
-
Chromosome Location:
Chr.17: 8929675 - 8980712 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01448619_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcbd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003751642.2 XP_003751690.1 3 - 4 435 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

tetrahydrobiopterin biosynthetic process
positive regulation of transcription, DNA-templated
protein homotetramerization
protein heterooligomerization
oxidation-reduction process
phenylalanine 4-monooxygenase activity
4-alpha-hydroxytetrahydrobiopterin dehydratase activity

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054743 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054743 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054743 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1379329_PM_at - -
Rat Genome 230 2.0 Array 1379329_at - -
Rat Genome 230 2.0 Array 1379330_PM_s_at - -
Rat Genome 230 2.0 Array 1379330_s_at - -
Affymetrix HT Rat Focus Array 1379330_PM_s_at - -
Affymetrix HT Rat Focus Array 1379330_s_at - -
Affymetrix HT Rat Focus Array 1379329_PM_at - -
Affymetrix HT Rat Focus Array 1379329_at - -
Rat Expression Set 230 1379329_PM_at - -
Rat Expression Set 230 1379330_s_at - -
Rat Expression Set 230 1379330_PM_s_at - -
Rat Expression Set 230 1379329_at - -
Rat Genome U34 Set rc_AI044449_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21508962 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0