This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01450153_m1
See other NUDT9 GE Assays ›
Gene Symbol:
Nudt9
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 9
Gene Aliases:
-
Chromosome Location:
Chr.14: 7036816 - 7054659 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01450153_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001006991.1 NP_001006992.1 7 - 8 966 100
XM_006250630.2 XP_006250692.1 6 - 7 767 100
XM_006250631.2 XP_006250693.1 7 - 8 789 100
GenBank mRNA BC083722.1 - 7 - 8 966 7190984 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1400001329.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nudt9

Panther Classification:

Molecular Function -

hydrolase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

ADP catabolic process
IDP catabolic process
adenosine-diphosphatase activity
ADP-ribose diphosphatase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P386799 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 876811 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 248624 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 489092 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 594982 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 716639 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 502784 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 187275 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 106284 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 521252 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 853332 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 476962 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 443313 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 708134 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 379777 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 353468 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 68264 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 47452 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P386799 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P386799 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P386799 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P386799 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 27717 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 59688 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10775403 - -
Affymetrix Rat Gene 1.0 ST Array 10775402 - -
Affymetrix Rat Gene 1.0 ST Array 10775400 - -
Affymetrix Rat Gene 1.0 ST Array 10775404 - -
Affymetrix Rat Gene 1.0 ST Array 10775401 - -
Affymetrix Rat Gene 1.0 ST Array 10775405 - -
Affymetrix Rat Gene 1.0 ST Array 10775399 - -
Affymetrix Rat Gene 1.0 ST Array 10775406 - -
Affymetrix Rat Gene 1.1 ST Array 10775398 - -
Rat Genome 230 2.0 Array 1373840_PM_at - -
Rat Genome 230 2.0 Array 1373840_at - -
Affymetrix HT Rat Focus Array 1373840_at - -
Affymetrix HT Rat Focus Array 1373840_PM_at - -
Affymetrix Rat Clariom S Assay PSR1400009953.rn.1 TC1400001329.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400009954.rn.1 TC1400001329.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400009957.rn.1 TC1400001329.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400009955.rn.1 TC1400001329.rn.1 RTA TC
Rat Expression Set 230 1373840_PM_at - -
Rat Expression Set 230 1373840_at - -
Affymetrix Rat Gene 1.0 ST Array 10775398 - -
Rat Genome U34 Set rc_AI179709_at - -
Rat Genome U34 Set rc_AI104864_at - -
Rat Genome U34 Set rc_AI105432_at - -
Rat Genome U34 Set rc_AI104864_g_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21341740 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1349382 - -
RatRef-12 Expression BeadChip ILMN_68386 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0