This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01454496_g1
See other EEF1G GE Assays ›
Gene Symbol:
Eef1g
Gene Name:
eukaryotic translation elongation factor 1 gamma
Gene Aliases:
-
Chromosome Location:
Chr.1: 225163391 - 225173997 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01454496_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef1g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004223.2 NP_001004223.1 6 - 7 855 117
GenBank mRNA BC079398.1 - 7 - 8 864 7112627 117
BC098895.1 - 6 - 7 855 7367246 117
FQ212596.1 - 7 - 8 873 117
FQ213179.1 - 7 - 8 872 117
FQ214210.1 - 7 - 8 870 117
FQ214443.1 - 7 - 8 873 117
FQ215853.1 - 7 - 8 901 117
FQ216520.1 - 5 - 6 640 117
FQ218274.1 - 7 - 8 871 117
FQ219298.1 - 7 - 8 873 117
FQ222930.1 - 7 - 8 872 117
FQ223614.1 - 7 - 8 871 117
FQ224956.1 - 6 - 7 831 117
FQ226853.1 - 4 - 5 544 117
FQ228703.1 - 7 - 8 871 117
FQ228918.1 - 7 - 8 873 117
FQ230995.1 - 7 - 8 871 117
FQ231923.1 - 7 - 8 873 117
FQ233868.1 - 7 - 8 871 117
FQ234035.1 - 7 - 8 871 117
FQ235034.1 - 7 - 8 1023 117

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

translational elongation
response to virus
translation elongation factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100026166.rn.1 TC0100003167.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0