This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01454645_m1
See other LIPN GE Assays ›
Gene Symbol:
Lipn
Gene Name:
lipase, family member N
Gene Aliases:
RGD1560354
Chromosome Location:
Chr.1: 252377169 - 252393340 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01454645_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001191955.1 NP_001178884.1 4 - 5 529 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100003507.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
hydrolase activity, acting on ester bonds

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100028414.rn.1 TC0100003507.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100028411.rn.1 TC0100003507.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100028413.rn.1 TC0100003507.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100028416.rn.1 TC0100003507.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100028415.rn.1 TC0100003507.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0