This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01454654_m1
See other MS4A6A GE Assays ›
Gene Symbol:
Ms4a6a
Gene Name:
membrane-spanning 4-domains, subfamily A, member 6A
Gene Aliases:
Ms4a11
Chromosome Location:
Chr.1: 227919284 - 227932637 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01454654_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ms4a6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_342028.7 XP_342029.5 6 - 7 816 134

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

transporter

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P993328 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P993328 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P993328 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P993328 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P993328 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P384404 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1376390_at - -
Rat Genome 230 2.0 Array 1392819_at - -
Rat Genome 230 2.0 Array 1392819_PM_at - -
Rat Genome 230 2.0 Array 1376390_PM_at - -
Affymetrix HT Rat Focus Array 1392819_at - -
Affymetrix HT Rat Focus Array 1392819_PM_at - -
Affymetrix HT Rat Focus Array 1376390_at - -
Affymetrix HT Rat Focus Array 1376390_PM_at - -
Rat Expression Set 230 1376390_at - -
Rat Expression Set 230 1376390_PM_at - -
Rat Expression Set 230 1392819_at - -
Rat Expression Set 230 1392819_PM_at - -
Rat Genome U34 Set rc_AA963068_at - -
Rat Genome U34 Set rc_AI009669_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21572951 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1356010 - -
RatRef-12 Expression BeadChip ILMN_64075 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0