This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01454704_m1
See other ETFB GE Assays ›
Gene Symbol:
Etfb
Gene Name:
electron-transfer-flavoprotein, beta polypeptide
Gene Aliases:
-
Chromosome Location:
Chr.1: 98472776 - 98486940 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01454704_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Etfb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004220.1 NP_001004220.1 3 - 4 401 100
GenBank mRNA BC079351.1 - 3 - 4 401 7108112 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100008117.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Etfb

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
oxidation-reduction process
molecular_function
electron carrier activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100066625.rn.1 TC0100008117.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0