This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01456648_m1
See other SMG8 GE Assays ›
Gene Symbol:
Smg8
Gene Name:
SMG8 nonsense mediated mRNA decay factor
Gene Aliases:
RGD1306862
Chromosome Location:
Chr.10: 74379224 - 74388846 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01456648_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smg8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_213430.7 XP_213430.2 1 - 2 2372 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
regulation of protein kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline