This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01456770_m1
See other SPNS3 GE Assays ›
Gene Symbol:
Spns3
Gene Name:
spinster homolog 3 (Drosophila)
Gene Aliases:
RGD1561884
Chromosome Location:
Chr.10: 59056460 - 59113312 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01456770_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spns3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109035.1 NP_001102505.1 2 - 3 437 59
XM_006246786.2 XP_006246848.1 3 - 4 1026 59
XM_006246789.2 XP_006246851.1 3 - 4 1027 59
XM_008767857.1 XP_008766079.1 3 - 4 1027 59
XM_008767858.1 XP_008766080.1 3 - 4 1027 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000003181.rn.1

Set Membership:

Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

sphingosine-1-phosphate signaling pathway
lipid transport
locomotion
transmembrane transport
sphingolipid transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000028713.rn.1 TC1000003181.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7xkh5:80/100.66.78.74:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0